missing translation for 'onlineSavingsMsg'
Learn More

Cefsulodin Sodium Salt, TRC

Product Code. 30134923
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134923 1 g 1g
30133516 5 g 5g
17974665 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134923 Supplier Toronto Research Chemicals Supplier No. TRCC2480901G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52152-93-9
Molecular Formula C22 H19 N4 O8 S2 . Na
Molecular Weight (g/mol) 554.53
Synonym Pyridinium, 4-(aminocarbonyl)-1-[[(6R,7R)-2-carboxy-8-oxo-7-[[(2R)-2-phenyl-2-sulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-, inner salt, sodium salt (1:1), Pyridinium, 4-(aminocarbonyl)-1-[[(6R,7R)-2-carboxy-8-oxo-7-[[(2R)-phenylsulfoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-, inner salt, monosodium salt (9CI), Pyridinium, 4-(aminocarbonyl)-1-[[2-carboxy-8-oxo-7-[(phenylsulfoacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-, hydroxide, inner salt, monosodium salt, [6R-[6α,7β(R*)]]-, 7-(D-α-Sulfophenylacetamido)ceph-3-em-3-(4'-carbamoylpyridinium)methyl-4-carboxylate sodium salt, Abbot 46811, CGP 7174E, Cefomonil, Cefsulodin sodium, D-(-)-SCE-129, Monaspor, Monosodium cefsulodin, Pseudocef, Pseudomonil, Pyocefal, Pyridinium, 4-(aminocarbonyl)-1-[[2-carboxy-8-oxo-7-[(phenylsulfoacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-, inner salt, monosodium salt, [6R-[6α,7β(R*)]]-, SCE 129, Sodium cefsulodin, Sulcephalosporin, Takesulin, Tilmapor, Ulfar, Cefsulodin Sodium
IUPAC Name sodium;(6R,7R)-3-[(4-carbamoylpyridin-1-ium-1-yl)methyl]-8-oxo-7-[[(2R)-2-phenyl-2-sulfonatoacetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES [Na+].NC(=O)c1cc[n+](CC2=C(N3[C@H](SC2)[C@H](NC(=O)[C@@H](c4ccccc4)S(=O)(=O)[O-])C3=O)C(=O)[O-])cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C22H20N4O8S2.Na/c23-18(27)13-6-8-25(9-7-13)10-14-11-35-21-15(20(29)26(21)16(14)22(30)31)24-19(28)17(36(32,33)34)12-4-2-1-3-5-12;/h1-9,15,17,21H,10-11H2,(H4-,23,24,27,28,30,31,32,33,34);/q;+1/p-1/t15-,17-,21-;/m1./s1
Formula Weight 554.0542
Chemical Name or Material Cefsulodin Sodium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.