missing translation for 'onlineSavingsMsg'
Learn More

Ceftazidime-d5 (>90%), TRC

Product Code. 30362142
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 μg
Unit Size:
10mg
1mg
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362142 50 μg Pack of 50
30356553 10 mg 10mg
30351383 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30362142 Supplier Toronto Research Chemicals Supplier No. TRCC24410250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ceftazidime-d5
Molecular Formula C22H17D5N6O7S2
Quantity 50 μg
Synonym 1-[[(6R,7R)-7-[[(2Z)-2-(2-Amino-4-thiazolyl)-2-[(1-carboxy-1-methylethoxy)imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]pyridine-d5, Biotum-d5, Ceftacidin-d5, Fortam-d5, Fortaz-d5, Fortum-d5, GR 20263-d5, Modacin-d5, Tazicef-d5, Tazidime-d5
InChI Formula InChI=1S/C22H22N6O7S2/c1-22(2,20(33)34)35-26-13(12-10-37-21(23)24-12)16(29)25-14-17(30)28-15(19(31)32)11(9-36-18(14)28)8-27-6-4-3-5-7-27/h3-7,10,14,18H,8-9H2,1-2H3,(H4-,23,24,25,29,31,32,33,34)/b26-13+/t14-,18-/m1/s1/i3D,4D,5D,6D,7D
SMILES [H][C@@]1([C@]([H])(NC(/C(C2=CSC(N)=N2)=N\OC(C)(C)C(O)=O)=O)C3=O)N3C(C([O-])=O)=C(C[N+]4=C([2H])C([2H])=C([2H])C([2H])=C4[2H])CS1
IUPAC Name (6R,7R)-7-[[(2E)-2-(2-amino-1,3-thiazol-4-yl)-2-(2-carboxypropan-2-yloxyimino)acetyl]amino]-8-oxo-3-[(2,3,4,5,6-pentadeuteriopyridin-1-ium-1-yl)methyl]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Molecular Weight (g/mol) 551.61
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.