missing translation for 'onlineSavingsMsg'
Learn More

Ceftazidime pentahydrate, 98%, Thermo Scientific Chemicals

Product Code. 15780909 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15780909 5 g 5g
15741767 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15780909 Supplier Thermo Scientific Acros Supplier No. 461730050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78439-06-2
Molecular Formula C22H22N6O7S2·5H2O
Molecular Weight (g/mol) 636.65
InChI Key NMVPEQXCMGEDNH-TZVUEUGBSA-N
PubChem CID 131673973
ChEBI CHEBI:3509
IUPAC Name 2-[(Z)-[1-(2-amino-1,3-thiazol-4-yl)-2-[[(6R,7R)-2-carboxy-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-en-7-yl]amino]-2-oxoethylidene]amino]oxy-2-methylpropanoate;pentahydrate
SMILES CC(C)(C(=O)[O-])ON=C(C1=CSC(=N1)N)C(=O)NC2C3N(C2=O)C(=C(CS3)C[N+]4=CC=CC=C4)C(=O)O.O.O.O.O.O

Specifications

Melting Point >150.0°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Merck Index 13,1959
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alkali,slightly soluble in methanol,dmf,insoluble in acetone,ethanol,chloroform,ether,,ethyl acetate,toluene,dioxane
Formula Weight 636.65
Percent Purity 98%
Chemical Name or Material Ceftazidime pentahydrate

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.