missing translation for 'onlineSavingsMsg'
Learn More

Ceftazidime t-Butyl Ester, TRC

Product Code. 30352937
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30352937 5 mg 5mg
30350627 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30352937 Supplier Toronto Research Chemicals Supplier No. TRCC2441155MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102772-66-7
Molecular Formula C26 H30 N6 O7 S2
Molecular Weight (g/mol) 602.68
Synonym Pyridinium, 1-[[(6R,7R)-7-[[(2Z)-2-(2-amino-4-thiazolyl)-2-[[2-(1,1-dimethylethoxy)-1,1-dimethyl-2-oxoethoxy]imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-, inner salt, Pyridinium, 1-[[(6R,7R)-7-[[(2Z)-(2-amino-4-thiazolyl)[[2-(1,1-dimethylethoxy)-1,1-dimethyl-2-oxoethoxy]imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-, inner salt (9CI), Pyridinium, 1-[[7-[[(2-amino-4-thiazolyl)[[2-(1,1-dimethylethoxy)-1,1-dimethyl-2-oxoethoxy]imino]acetyl]amino]-2-carboxy-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-, inner salt, [6R-[6α,7β(Z)]]-
IUPAC Name (6R,7R)-7-[[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-[2-methyl-1-[(2-methylpropan-2-yl)oxy]-1-oxopropan-2-yl]oxyiminoacetyl]amino]-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
SMILES CC(C)(C)OC(=O)C(C)(C)O\N=C(/C(=O)N[C@H]1[C@H]2SCC(=C(N2C1=O)C(=O)[O-])C[n+]3ccccc3)\c4csc(N)n4

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C26H30N6O7S2/c1-25(2,3)38-23(37)26(4,5)39-30-16(15-13-41-24(27)28-15)19(33)29-17-20(34)32-18(22(35)36)14(12-40-21(17)32)11-31-9-7-6-8-10-31/h6-10,13,17,21H,11-12H2,1-5H3,(H3-,27,28,29,33,35,36)/b30-16-/t17-,21-/m1/s1
Formula Weight 602.1617
Chemical Name or Material Ceftazidime t-Butyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.