missing translation for 'onlineSavingsMsg'
Learn More

Celastrol (>85%), TRC

Product Code. 30355661
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity unitSize
30355661 50 mg 50mg
1 options
This item is not returnable. View return policy

Product Code. 30355661

Brand: Toronto Research Chemicals TRCC24951050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Celastrol
CAS 34157-83-0
Molecular Formula C29 H38 O4
Quantity 50 mg
Synonym 24,25,26-Trinoroleana-1(10),3,5,7-tetraen-29-oic acid, 3-hydroxy-9,13-dimethyl-2-oxo-, (9β,13α,14β,20α)-, 24-Nor-D:A-friedooleana-1(10),3,5,7-tetraen-29-oic acid, 3-hydroxy-2-oxo- (7CI,8CI), D:A-Friedo-24-noroleana-1(10),3,5,7-tetraen-29-oic acid, 3-hydroxy-2-oxo-, (20α)-, (9β,13α,14β,20α)-3-Hydroxy-9,13-dimethyl-2-oxo-24,25,26-trinoroleana-1(10),3,5,7-tetraen-29-oic acid, 2-Picenecarboxylic acid, 1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydro-10-hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-, (2R,4aS,6aS,12bR,14aS,14bR)-, 2-Picenecarboxylic acid, 1,2,3,4,4a,5,6,6a,11,12b,13,14,14a,14b-tetradecahydro-10-hydroxy-2,4a,6a,9,12b,14a-hexamethyl-11-oxo-, [2R-(2α,4aβ,6aβ,12bβ,14aα,14bβ)]-, Celastrol, NSC 70931, Tripterin
InChI Formula InChI=1S/C29H38O4/c1-17-18-7-8-21-27(4,19(18)15-20(30)23(17)31)12-14-29(6)22-16-26(3,24(32)33)10-9-25(22,2)11-13-28(21,29)5/h7-8,15,22,31H,9-14,16H2,1-6H3,(H,32,33)/t22-,25-,26-,27+,28-,29+/m1/s1
SMILES C[C@@]12[C@]([C@@]3([H])[C@@](CC1)(CC[C@](C)(C3)C(O)=O)C)(CC[C@@]4(C2=CC=C5C4=CC(C(O)=C5C)=O)C)C
IUPAC Name (2R,4aS,6aR,6aS,14aS,14bR)-10-hydroxy-2,4a,6a,6a,9,14a-hexamethyl-11-oxo-1,3,4,5,6,13,14,14b-octahydropicene-2-carboxylic acid
Molecular Weight (g/mol) 450.61
Formula Weight 450.277
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.