missing translation for 'onlineSavingsMsg'
Learn More

Celiprolol hydrochloride

Product Code. 15447139
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15447139 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15447139 Supplier Thermo Scientific Alfa Aesar Supplier No. J65943.MA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 57470-78-7 | C20H34ClN3O4 | 415.96 g/mol

A cardioselective 1-adrenergic blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 57470-78-7
Color White
Molecular Formula C20H34ClN3O4
MDL Number MFCD00941500
Quantity 10 mg
Merck Index 14,1960
Synonym celiprolol hydrochloride, celiprolol hcl, selectrol, selecor, selectol tn, celectol, ccris 1095, celiprolol hydrochloride usan, celiprolol clorhidrato spanish, celiprolol hydrochlorid german
Solubility Information Soluble in ethanol, methanol and water
InChI Key VKJHTUVLJYWAEY-UHFFFAOYNA-N
SMILES [H+].[Cl-].CCN(CC)C(=O)NC1=CC=C(OCC(O)CNC(C)(C)C)C(=C1)C(C)=O
IUPAC Name 3-[3-acetyl-4-[3-(tert-butylamino)-2-hydroxypropoxy]phenyl]-1,1-diethylurea;hydrochloride
Molecular Weight (g/mol) 415.96
PubChem CID 42373
Formula Weight 415.95
Physical Form Crystalline Solid
Chemical Name or Material Celiprolol hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.