missing translation for 'onlineSavingsMsg'
Learn More

Celite™ 535 Calcined Filter Aid, Honeywell Fluka™

Product Code. 15662120 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15662120 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15662120 Supplier Honeywell-Fluka Supplier No. 221381KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Filter aid, flux calcined

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Color Beige to White
Quantity 1 kg
CAS 68855-54-9
Packaging Fiberboard box
Molecular Formula CNa2O3
MDL Number MFCD00132803
UN Number NONH for all modes of transport
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES C(=O)([O-])[O-].[Na+].[Na+]
IUPAC Name disodium;carbonate
Molecular Weight (g/mol) 105.988
PubChem CID 10340
ChEBI CHEBI:29377
Chemical Name or Material Diatomaceous Earth, Flux-Calcined, Flux calcined filter aid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.