missing translation for 'onlineSavingsMsg'
Learn More

Celite™ 577 Fine, Solstice

Product Code. 15642130
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15642130 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15642130 Supplier Honeywell-Fluka Supplier No. 221421KG
  • ONLINE EXCLUSIVE PRICE
    95.00 €
    88.90 € / 1kg
    Save 6.10 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Filter aid, calcined

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Color White
Quantity 1 kg
CAS 91053-39-3
Packaging Drum with Inliner
Molecular Formula O2Si
MDL Number MFCD00132803
UN Number NONH for all modes of transport
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name dioxosilane
Molecular Weight (g/mol) 60.083
PubChem CID 24261
ChEBI CHEBI:30563
Chemical Name or Material Diatomaceous Earth, Calcined, Calcined
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.