missing translation for 'onlineSavingsMsg'
Learn More

Celite™, for analysis, high purity analytical grade

Product Code. 10295890 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass Bottle
Unit Size:
10g
25g
This item is not returnable. View return policy

Product Code. 10295890

Brand: Thermo Scientific Acros 173460250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 61790-53-2 | O2Si | 60.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61790-53-2
Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00011232
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym Infusorial earth, Diatomite, Diatomaceous earth
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Physical Form Powder
Density <0.25 g/mL (centrifuged wet density)
Color Gray/Pink to White
Quantity 25 g
pH 5 to 8 (25°C, 10% aq. suspension)
Loss on Drying 0.5% max.
Loss on Ignition 2% max.
Packaging Glass Bottle
Permeability 0.15 to 0.30 DARCY
Silicon Dioxide 98.7%
Formula Weight 60.08
Grade Analytical
Chemical Name or Material Celite™
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.