missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Cell counting kit-8

Product Code. 18725765
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18725765 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18725765 Supplier R&D Systems™ Supplier No. 7368/5ML
  • ONLINE EXCLUSIVE PRICE
    266.00 €
    232.05 € / 5mL
    Save 33.95 €
    12% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cell viability and proliferation assay test solution

Cell Counting Kit-8 is a viability and proliferation assay test solution. It is a supplied as a ready-to-use 1-bottle solution and enables the colorimetric determination of the number of viable cells in cell proliferation and cytotoxicity assays. Cell Counting Kit-8 utilizes a highly water-soluble tetrazolium salt (WST-8), which is reduced by cellular dehydrogenases to an orange formazan dye. The color change can be detected in the 430 - 490 nm range and Cell Counting Kit-8 exhibits improved sensitivity compared with other tetrazolium salt-based assays such as MTT, XTT or MTS. Can also be used in the detection of superoxide.

For more information on how to use Cell Counting Kit-8, please see our protocol .

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mL
Product Line Tocris
Target Cell Viability/Proliferation Assays
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.