Learn More
Cell Culture Grade Water Tested to USP/EP Sterile Water, Corning™
BIOPHARMA
Product Code. 15353661
Change view
Click to view available options
Quantity:
200 L
Unit Size:
200 L
Product Code. Quantity Unit Size
15353661 200 L 200 L
1 options
Product should not be used as an injectable or in the preparation of injectables.
Product Code. 15353661 Supplier Corning Media Tech Supplier No. 25065LH
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Product should not be used as an injectable or in the preparation of injectables.
High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP/EP Sterile Water are used in preparation of purification and chromatography buffers.
Cell culture grade water is high-quality water suitable for use as a solvent in the preparation of cell culture media and other reagents for sensitive applications. Our bioprocess container selections are suitable for large-scale applications such as cleaning and upstream and downstream reagent preparation. Cell culture grade water goes through a wide array of testing which includes testing to the chemical and physical properties found in the USP and EP monographs for sterile Water for Injection (WFI).
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
TRUSTED_SUSTAINABILITY
Specifications
Quantity 200 L