missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Cell Line Optimization 4D-Nucleofector™ X Kit

Product Code. 13439309
Change view
Click to view available options
No. of Reactions:
24
96
Unit Size:
24 reactions
96 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
13439309 24 24 reactions
13409339 96 96 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13439309 Supplier Lonza™ Supplier No. V4XC1024
  • ONLINE EXCLUSIVE PRICE
    810.00 €
    674.78 € / 24 reactions
    Save 135.22 €
    16% Off
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Content And Storage 4°C
Vector pmaxGFP™
For Use With (Equipment) 4D-Nucleofector™ X Unit
Includes Includes 100μL Transfection volume, 2.22mL Nucleofector Solution, 0.5mL Supplement, 50μg pmaxGFP Vector, 24 Single Nucleocuvette (100 μl), Store at 4°C, For Use with 4D-Nucleofector X Unit
No. of Reactions 24
Product Type Cell Line Optimization 4D-Nucleofector X Kit
Quantity 1 Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.