Learn More
TPP™ Rotatable Cell Scrapers
Product Code. 11371714
Change view
Click to view available options
Length (Metric) Handle:
240 mm
Unit Size:
Pack of 150
Product Code. Length (Metric) Handle Unit Size
11371714 240 mm Pack of 150
1 options
Product Code. 11371714 Supplier TPP™ Supplier No. 99002
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Designed with sharp edge blade to collect clean and complete cell sample. TPP™ Rotatable Cell Scrapers feature a free rotating blade to provide access to all corners.
To change blade angle: apply slight pressure pushing the handle downward towards the floor of the container; slight rotation of the handle will cause the blade to twist to the desired direction.
  • Small raised knobs on the handle for a slip free and secure grip with rubber gloves
  • Lint-free blister packaging
  • Single wrapped, lint-free paper/plastick blister packaging
  • Convenient, stackable dispenser box 
Specifications
Width (Metric) Blade 13 mm
For Use With (Equipment) Tissue culture flasks, flat tubes, dishes
Length (Metric) Handle 240 mm
Material Polyethylene