missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellBlox™ Plus Blocking Buffer

Product Code. 17952031
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
500 Tests
Unit Size:
Pack of 100
Pack of 25
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952031 25 Tests Pack of 25
17912071 100 Tests Pack of 100
17912061 500 Tests Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17952031 Supplier Invitrogen™ Supplier No. C001T02F01
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Invitrogen CellBlox Plus Blocking Buffer is formulated to block nonspecific binding of NovaFluor labels with cells.

Invitrogen CellBlox Plus Blocking Buffer is formulated to block nonspecific binding of NovaFluor labels with cells. These nonspecific interactions can result in higher background labeling. CellBlox Plus Blocking Buffer is a non-antibody blocking solution and should be used whenever a NovaFluor dye is used for labeling of any cell type. CellBlox Plus Blocking Buffer has enhanced blocking capabilities compared to our original CellBlox Blocking Buffer (Figure 1).

CellBlox Plus Blocking Buffer is also recommended for use with cyanine-based dyes or cyanine-based tandem dyes to block non-specific interactions with monocytes and macrophages to minimize background labeling (Figure 2).

Features of CellBlox Plus Blocking Buffer include:
• Compatible with all fluorophores and with LIVE/DEAD Fixable dead cell stains
• Can be used with any fluorophore-antibody conjugate as high-performance monocyte and macrophage blocking solution
• Compatible with other blocking reagents, such as Fc Block, blocking proteins, Brilliant Stain Buffer, and Super Bright Complete Staining Buffer
• Not required when labeling antibody-capture beads, such as UltraComp eBeads Plus Compensation Beads

Use of CellBlox Plus Blocking Buffer requires minimal change to most flow cytometry staining protocols. Simply add CellBlox Plus Blocking Buffer to an antibody cocktail mixture prior to labeling cells. Specifically, add 5 μL CellBlox Plus Blocking Buffer for every stained sample to be labeled with the antibody cocktail mixture, with 100 μL as a final staining volume.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Flow Cytometry
For Use With (Equipment) Flow Cytometer
Physical Form Liquid
Product Line eBioscience
Quantity 25 Tests
Solution Type Buffer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.