Learn More
Corning™ CellCube™ Modules, With Aseptic Connectors
Shop All Corning Products
Click to view available options
Tray Layers:
10-Layer
100-Layer
25-Layer
Unit Size:
Each
Pack of 2
Description
- Uses a tissue culture-treated growth surface for cell attachment, and continually perfuses the cells with fresh medium for increased cell productivity
- The CellCube system provides an environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen with low differential gradients across all cells within the modules.
- The CellCube modules provide a traditional tissue culture-treated surface or Corning CellBIND™ surface for the growth of attachment dependent cells.
Specifications
Specifications
| Includes | Growth Surface and Aseptic Connectors |
| No. per Case | 1/Cs. |
| No. per Pack | 1/Pk. |
| Sterility | Sterile |
| Culture Area | 85,000 cm² |
| Tray Layers | 100-Layer |