missing translation for 'onlineSavingsMsg'
Learn More

AcroBiosystems Cellectra Human CD14 nBeads, premium grade (for cells)

Product Code. 31451885 Shop All ACROBiosystems Products
Change view
Click to view available options
Quantity:
1 x 109 cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31451885 1 x 109 cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31451885 Supplier AcroBiosystems Supplier No. MBSS0011×10^9cells
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cellectra Human CD14 nBeads, premium grade (for cells) is expressed from Human HEK293.

Cellectra Human CD14 nBeads, premium grade (for cells) is expressed from Human HEK293.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This product is stable after storage at: 2°C to 8°C for 36 months in lyophilized state; 2°C to 8°C for 18 months under sterile conditions after reconstitution. Please avoid repeated freeze-thaw cycles once reconstituion.
Particle Size 50 nm
Form Lyophilized
Quantity 1 x 109 cells
For Use With (Application) For the positive selection of monocytes and macrophages from fresh or frozen human peripheral blood mononuclear cells (PBMCs)
Species Human
Sufficient For 1 x 109 cells
Regulatory Status RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.