missing translation for 'onlineSavingsMsg'
Learn More

Molecular Biology Grade Water Tested to USP Sterile water, Corning™
BIOPHARMA

Product Code. 15393581
Change view
Click to view available options
Quantity:
6 x 100 mL
6 x 500 mL
6 x 1 L
Unit Size:
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15393581 6 x 500 mL Pack of 6
15383581 6 x 100 mL Pack of 6
15303711 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15393581 Supplier Corning Media Tech Supplier No. 46000CV
  • ONLINE EXCLUSIVE PRICE
    158.00 €
    136.61 € / Pack of 6
    Save 21.39 €
    13% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in RNase, DNase and Protease free applications. Corning™ Molecular Biology Grade Water Tested to USP Sterile water is used for Buffer preparation, DNA/RNA purification, DNA sequencing and libraries, recombinant DNA, cloning, gene mapping.

Molecular biology grade water is ideal for applications where highly pure, RNase-, DNase-, and protease-free water is required (e.g., PCR and other molecular biology reactions setup, DNA/RNA/protein extraction, purification, and storage reagents). Molecular biology grade water is tested to the chemical and physical properties found in the USP monograph for sterile purified water.

Certifications

Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Quantity 6 x 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.