missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CellInsight™ CX7 High Content Analysis Platform and Store Standard Edition

Product Code. 15910603 Shop All Thermo Scientific Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
15910603 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For research use only
Product Code. 15910603 Supplier Thermo Scientific™ Supplier No. CX7B1112

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy
For research use only

A fast, automated cellular imaging and analysis platform designed for quantitative microscopy.

Integrated system for high content analysis

The Thermo Scientific™ CellInsight™ CX7 High Content Analysis (HCA) Platform is a fast, automated cellular imaging and analysis platform designed for quantitative microscopy.

The CellInsight CX7 HCA Platform offers an integrated way to develop and automate high content assays using a combination of 5-color brightfield and 7-color fluorescence imaging in confocal or wide-field modes. See how the CellInsight CX7 platform compares to other high content analysis platforms from Thermo Fisher Scientific.

Features of the CellInsight CX7 HCA Platform
  • 7-channel fluorescent imaging
  • 5-channel bright-field imaging
  • 7-channel confocal imaging
  • Software and laser-based autofocus for consistent scan times
  • Configured for fully automated plate handling and scanning

Capabilities
  • Addresses hundreds of assay types with flexible software and hardware tools
  • Scan and analyze microscope slides or most SBS-standard format microplates
  • Use HCS Studio software for integrated instrument control and data analysis
  • Achieve the fastest time to results by using intelligent acQuisition (iQ) real-time analysis feedback
  • Compatible with Molecular Probes™ fluorescent detection reagents
Hardware and Software
  • Stable and reliable light source enables reduced variability, increased data quality, and decreased operating costs
  • Broad reagent options with seven-color excitation from violet to near IR for assay multiplexing
  • Dual mode focusing using Software and Laser Autofocus methods to address both labelled and unlabeled sample types
  • Multi-mode imaging allows for wide-field, bright-field, and confocal imaging in one run
  • HCS Studio Cell Analysis Software is an integrated instrument control, visualization, and analysis software package for novice to advanced users
  • Scalable with Store Image and Database Software and optional Orbitor RS Microplate handler
TRUSTED_SUSTAINABILITY

Specifications

Product Line CellInsight
Description CellInsight™ CX7 High Content Analysis Platform and Store Standard Edition
For Use With (Equipment) High Content Instrument
Color Assorted Colors
Detection Method Chromogenic, Fluorescence
Format Microplate, Slide(s)
Content And Storage • 1 Reader
• 1 Store Software
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.