missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CellLight™ Actin-GFP, BacMam 2.0

Product Code. 10110979
Change view
Click to view available options
Quantity:
100μL
1 mL
Target:
Actin
Actin, Cytoskeleton
Unit Size:
100µL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Target unitSize
10110979 1 mL Actin, Cytoskeleton Each
13276509 100μL Actin 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10110979 Supplier Invitrogen™ Supplier No. C10582

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Provides easy method for labeling of actin with Green Fluorescent Protein in live cells

This ready-to-use construct is transduced/transfected into cells using BacMam 2.0 technology, where it expresses GFP fused to human actin. You can observe actin-GFP behavior in live cells using a fluorescent imaging system and standard FITC/GFP filter set. Study dynamic action of actin filaments in live cells, with almost no cytotoxicity. In addition, you can co-transduce/transfect more than one CellLight reagent and label live cells with multiple tracking or tracing dyes to image dynamic cellular processes. Cells expressing CellLight constructs can also be fixed with formaldehyde for multiplexing with antibodies using immunocytochemical techniques.

BacMam Technology

CellLight Actin-GFP, BacMam 2.0, is a fusion construct of human actin and emGFP, providing accurate and specific targeting to cellular actin-GFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.

  • Highly efficient: up to 90% transduction of a wide range of mammalian cell lines, including primary cells, stem cells, and neurons
  • Fast and convenient: simply add CellLight reagent to cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
  • Less toxic: CellLight reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling
  • Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose
  • Flow Cytometer Laser Lines: 488

Cell Analysis, Cell Structure, Cell Viability, Proliferation & Function, Cellular Imaging, Chemotaxis & Cell Migration, Cytoskeleton, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining & Detection

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage Store at 2°C to 6°C, protected from light. Do Not Freeze.
Excitation Wavelength Range 488⁄510
Dye Type GFP (EmGFP)
For Use With (Equipment) Confocal Microscope, Fluorescence Microscope
Quantity 1 mL
Detection Method Fluorescence
Emission Visible
Form Liquid
Product Line CellLight
Shipping Condition Wet Ice
Target Actin, Cytoskeleton
Technique Fluorescence Intensity
Label Type Fluorescent Dye
Product Type Actin
SubCellular Localization Actin, Cytoskeleton
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.