missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CellSensor™ AP-1-bla A375 Cell Line

Product Code. 10732924
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10732924 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10732924 Supplier Thermo Scientific™ Supplier No. K1660
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The CellSensor™ AP1-bla A375 cell line contains a beta-lactamase reporter gene under control of the AP1 response element stably integrated into A375 cells.

The CellSensor™ AP1-bla A375 cell line contains a beta-lactamase reporter gene under control of the AP1 response element stably integrated into A375 cells. A375 cells are human melanoma cancer cells that contain the endogenous B-Raf mutation V600E resulting in constitutive B-Raf kinase activity. The CellSensor™ AP1-bla A375 cell line is a clonal population isolated by flow cytometry based on constitutive expression of beta-lactamase.

This cell line has been validated with various small-molecule Raf inhibitors as well as B-Raf Stealth RNAi™ siRNA. This cell line has also been tested for assay performance under variable conditions, including DMSO concentration, cell number, compound incubation time, and substrate loading time and validated for Zfi-factor and IC50 concentrations of Raf1 Inhibitor I. The RAF gene family (RAF1, A-RAF, and B-RAF) encodes closely related serine/threonine protein kinases that are important effectors of Ras activation. Raf1 and A-Raf are rarely mutated, whereas mutations in B-Raf are common in human cancers, especially melanoma. B-Raf is mutated in about 70% of human melanomas, 35-70% of papillary thyroid carcinomas, and less commonly in lung and colorectal carcinomas. Mutations are mostly in the B-Raf kinase domain and, in melanomas, the vast majority are V600E missense mutations leading to activation of B-Raf kinase. The constitutive activity of B-Raf V600E can directly lead to the activation of the Mek/MAPK signaling pathway. Therefore, inhibition of B-Raf/Mek/MAPK signaling could be a potential way of treating melanomas and other tumors with mutant B-Raf.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
For Use With (Application) Signaling Pathway
Product Line CellSensor
Quantity 1 Vial
Target Entry MAPK pathway, BRAF V600E
Type Dividing Cells
System Type CellSensor™
Druggable Target Signaling Pathway

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.