missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CellSensor™ AP-1-bla HEK293T Cell Line

Product Code. 10062913
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10062913 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10062913 Supplier Thermo Scientific™ Supplier No. K1658
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The CellSensor™ AP-1-bla HEK 293T Cell Line contains a beta-lactamase reporter gene under control of the AP-1 response element stably integrated into HEK 293T cells.

The CellSensor™ AP-1-bla HEK 293T Cell Line contains a beta-lactamase reporter gene under control of the AP-1 response element stably integrated into HEK 293T cells. The cell line was created through sorting by FACS of cells responsive to stimulation of the AP-1 pathway with phorbol 12-myristate 13-acetate (PMA). The cell line was validated for DMSO tolerance, incubation time with stimulant, and substrate loading conditions. The AP-1-bla HEK 293T cell line responds to agonist treatment as expected from literature and can be adapted for high throughput screening for agonists or antagonists of the AP-1 pathway with compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Description CellSensor™ AP-1-bla HEK293T Cell Line, 1Vial, ∽2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, AP-1-bla HEK293T Cell Line, Fluorescent Detection Method, CellSensor™ Druggable Target, Adherent Growth Properties, Liquid Nitrogen Storage
Detection Method Fluorescent
For Use With (Application) Pharma and biopharma, drug discovery and development, target and lead identification and validation, pathway biology, cellular pathway analysis assays, cellsensor™ cellular pathway assays
Includes beta-lactamase reporter gene under control of the AP-1 response element stably integrated into HEK 293T cells
Product Line CellSensor
Quantity 1 Vial
Sample Type AP-1-bla HEK293T
Target Entry EGFR, ERK1/2, p38, MEK, MAPK pathway, c-MET, JNK, RON, RAF
Type Cell Line
System Type CellSensor™
Druggable Target Signaling Pathway
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.