Learn More
Thermo Scientific™ CellSensor™ CRE-bla CHO K1 Cell Line
Description
The CellSensor™ CRE-bla CHO-K1 cell line contains a beta-lactamase reporter gene under control of a cAMP response element (CRE) stably integrated into CHO-K1 cells. This cell line can be used as a parental cell line to build specific G-protein coupled receptor (GPCR) assays after transfection of additional genes of interest or to detect changes in intracellular cAMP levels. CRE-bla CHO-K1 cells have been shown to be responsive to forskolin stimulation. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Description | CellSensor™ CRE-bla CHO K1 Cell Line, 1Vial, ∽ 2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, CRE-bla CHO K1 Cell Line, Fluorescent Detection Method, CellSensor™ Druggable Target, Adherent Growth Properties, Liquid Nitrogen Storage |
| Detection Method | Fluorescent |
| For Use With (Application) | Pharma and biopharma, drug discovery and development, target and lead identification and validation, kinase biology, kinase pathway analysis, pathway biology, cellular pathway analysis assays, cellSensor™ cellular pathway assays |
| Includes | beta-lactamase (bla) reporter gene under control of a cAMP response element (CRE) stably integrated into CHO-K1 cells |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Sample Type | CRE-bla CHO K1 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.