missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CellSensor™ CRE-bla Jurkat Cell Line

Product Code. 10703684
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10703684 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10703684 Supplier Thermo Scientific™ Supplier No. K1635

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The CRE-bla Jurkat cell line contains a beta-lactamase reporter gene under control of a cAMP response element (CRE) stably integrated into Jurkat cells.

The CRE-bla Jurkat cell line contains a beta-lactamase reporter gene under control of a cAMP response element (CRE) stably integrated into Jurkat cells. This cell line can be used as a parental cell line to build specific G-protein coupled receptor (GPCR) assays after transfection of additional genes of interest or to detect changes in intracellular cAMP levels. CRE-bla Jurkat cells have been shown to be responsive to forskolin stimulation. Academic and non-profit customers, please inquire for special pricing.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Description CellSensor™ CRE-bla Jurkat Cell Line, 1Vial, ∽ 50,000,000 cells/vial at 50,000,000 cells/mL Quantity of Cells, CRE-bla Jurkat Cell Line, Fluorescent Detection Method, CellSensor™ Druggable Target, Suspension Growth Properties, Liquid Nitrogen Storage
Detection Method Fluorescent
For Use With (Application) Pharma and biopharma, drug discovery and development, target and lead identification and validation, pathway biology, cellular pathway analysis assays, cellsensor™ cellular pathway assays
Includes beta-lactamase reporter gene under control of a cAMP response element (CRE) stably integrated into Jurkat cells.
Product Line CellSensor
Quantity 1 Vial
Sample Type CRE-bla Jurkat
Target Entry PKA, cAMP, cAMP/PKA pathway
Type Cell Line
System Type CellSensor™
Druggable Target Signaling Pathway
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.