missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CellSensor™ irf1-bla TF-1 Cell Line

Product Code. 10052913
Change view
Click to view available options
Quantity:
1 Vial
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10052913 1 Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10052913 Supplier Thermo Scientific™ Supplier No. K1657

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Jak/Stat signaling pathways play essential roles in cellular responses to distinct cytokines. One of the Jak/Stat pathways, Jak2/Stat5, mediates cell proliferation in response to Interleukin-3 (IL-3), prolactin, erythropoietin (Epo), and granulocyte-macrophage colony stimulating factor (GM-CSF).

Jak/Stat signaling pathways play essential roles in cellular responses to distinct cytokines. One of the Jak/Stat pathways, Jak2/Stat5, mediates cell proliferation in response to Interleukin-3 (IL-3), prolactin, erythropoietin (Epo), and granulocyte-macrophage colony stimulating factor (GM-CSF). JAK2 gene knockout causes embryonic lethality due to defective erythropoiesis, suggesting that the Jak2/Stat5 pathway plays an important role in red blood cell formation.

The recent discovery of an activating mutation in JAK2 (V617F) present in a high percentage of myeloproliferative disease (MPD) patients suggests that the Jak2/Stat5 pathway may be a potential therapeutic target for certain forms of MPD. The activated Stat5 transcription factor recognizes and binds to a specific palindromic DNA sequence found in the promoter region of β-casein, interferon regulatory factor-1 (irf-1), and a number of other genes. The CellSensor™ irf1-bla TF1 Cell Line contains a beta-lactamase reporter gene under control of the irf-1 response element stably integrated into TF1 cells. TF1 cells are a human erythroleukemia cell line that is growth-dependent on GM-CSF and have an intact GM-CSF-JAK2-STAT5 pathway.

This cell line is validated for EC50 and Z'-factor using GM-CSF. This cell line has also been tested under variable experimental conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time. Responsiveness to EPO and Jak Inhibitor 1 was also tested (Figures 2 and 3). CellSensor™ irf1-bla TF1 cells were stimulated in triplicate with GM-CSF over the indicated concentration range in a 384-well format. Cells were incubated for 5 hr. with agonist and 0.5% DMSO and then combined with LiveBLAzer™-FRET B/G Substrate (CCF4-AM) for 2.5 hr. Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader. The 460/530 ratios were plotted for each replicate against the indicated concentrations of GM-CSF. CellSensor™ irf1-bla TF1 cells were stimulated with Epo over the indicated concentration range in a 384-well format.

The Response Ratios were plotted against the indicated concentrations of Epo. CellSensor™ irf1-bla TF1 cells were treated with Jak Inhibitor 1 over the indicated concentration range in a 384-well format. Cells were then stimulated with GM-CSF or EPO for 5 hr. in 0.5% DMSO and then combined with LiveBLAzer™-FRET B/G Substrate (CCF4 - AM) for 2.5 hr. Fluorescence emission values at 460 nm and 530 nm were obtained using a standard fluorescence plate reader. These values were converted to percent inhibition relative to a set of controls (unstimulated cells and EC80 of GM-CSF or EPO-treated cells) and plotted against the indicated concentrations of Jak Inhibitor 1.

Academic and non-profit customers, please inquire for special pricing.

TRUSTED_SUSTAINABILITY

Specifications

Assay Entry Cell-based beta lactamase reporter gene
Cell State Dividing Cells
Content And Storage CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
Description CellSensor™ irf1-bla TF-1 Cell Line, 1Vial, ∽ 40,000,000 cells/vial at 40,000,000 cells/mL Quantity of Cells, irf1-bla TF-1 Cell Line, Fluorescent Detection Method, CellSensor™ Druggable Target, Suspension Growth Properties, Liquid Nitrogen Storage
Detection Method Fluorescent
For Use With (Application) Pharma and biopharma, drug discovery and development, target and lead identification and validation, pathway biology, cellular pathway analysis assays, cellsensor™ cellular pathway assays
Includes beta-lactamase reporter gene under control of the interferon regulatory factor-1 (irf-1) response element stably integrated into TF-1 cells
Product Line CellSensor
Quantity 1 Vial
Sample Type irf1-bla TF-1
Target Entry JAK/STAT (GM-CSF, EPO, IL-3) pathway, JAK2, STAT5
Type Cell Line
Cell Line TF-1
System Type CellSensor™
Druggable Target Signaling Pathway
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.