Learn More
Thermo Scientific™ CellSensor™ ISRE-bla RA-1 Cell Line
Description
The CellSensor™ ISRE-bla RA-1 Cell Line contains the beta-lactamase (bla) gene under the control of the interferon-stimulated response element (ISRE). This cell line was engineered by lentiviral transduction of an ISRE-bla construct into RA-1 cells. Flow cytometry was then used to isolate a pool of cells responsive to polyinosinic-polycytidylic acid (poly I:C) stimulation. ISRE-bla RA-1 cells have been tested for assay performance using variable assay conditions, including DMSO concentration, cell number, stimulation time, substrate loading time, as well as validated for Z'-factor and EC50 concentrations of poly I:C. Additional testing data using alternate stimuli are also available. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Description | CellSensor™ ISRE-bla RA-1 Cell Line, 1Vial, ∽ 5,000,000 cells/vial at 5,000,000 cells/mL Quantity of Cells, ISRE-bla RA-1 Cell Line, Fluorescent Detection Method, CellSensor™ Druggable Target, Suspension Growth Properties, Liquid Nitrogen Storage |
| Detection Method | Fluorescent |
| For Use With (Application) | Pharma and biopharma, drug discovery and development, target and lead identification and validation, pathway biology, cellular pathway analysis assays, cellsensor™ cellular pathway assays |
| Includes | beta-lactamase gene under the control of the interferon stimulated response element (ISRE) |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Sample Type | ISRE-bla RA-1 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.