Learn More
Thermo Scientific™ CellSensor™ NFkB-bla RAW 264.7 Cell Line
Description
The CellSensor™ NFκB-bla RAW264.7 mouse macrophage cell line contains a beta-lactamase reporter gene under control of a stably integrated NFκB response element. This cell line is a clonal population isolated in response to lipopolysaccharide by flow cytometry. This cell line has been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, and substrate loading time, and has been validated for Z'-factor and EC50 concentrations of the TLR7 ligand imiquimod. Additional data using alternate stimuli are shown. The molecules responsible for coordinating the immune systems recognition of pathogens are Toll-like receptors (TLRs), of which there are currently 10 identified members in humans. TLRs are pattern recognition receptors (PRRs), which bind to pathogen-associated molecular patterns (PAMPs), small molecular sequences consistently found on pathogens. This binding activates the transcription of immune system genes and regulators such as cytokines, chemokines, and costimulatory molecules, thereby promoting the initial immune response of macrophages and neutrophils. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Description | CellSensor™ NFkB-bla RAW 264.7 Cell Line, 1Vial, ∽ 2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, NFkB-bla RAW 264.7 Cell Line, Fluorescent Detection Method, CellSensor™ Druggable Target, Adherent Growth Properties, Liquid Nitrogen Storage |
| Detection Method | Fluorescent |
| For Use With (Application) | Pharma and biopharma, drug discovery and development, target and lead identification and validation, pathway biology, cellular pathway analysis assays, cellsensor™ cellular pathway assays |
| Includes | beta-lactamase reporter gene under control of the NFkB response element that has been stably integrated into RAW 264.7 cells |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Sample Type | NFkB-bla RAW 264.7 |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.