missing translation for 'onlineSavingsMsg'
Learn More

CELLPATH CellSoft 4 x 125 mL

Product Code. 30226227
Change view
Click to view available options
Quantity:
4 x 125 mL
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30226227 4 x 125 mL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30226227 Supplier CELLPATH Supplier No. EAA150025A
  • ONLINE EXCLUSIVE PRICE
    136.00 €
    66.84 € / Pack of 4
    Save 69.16 €
    50% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cellsoft is developed to provide a high-performance tissue softening solution that reduces the hazard of risk for users and is particularly effective when used for sectioning hardened nail tissue.

  • Key Benefits:
  • Excellent for hard materials - improved sectioning and ribboning
  • Lower risk to users
  • Coloured and scented for clear identification
  • IVD CE marked - ensuring quality and consistency

Specifications

Dilution Ready to use
Product Type Tissue Softener
Quantity 4 x 125 mL
For Use With (Application) Tissue softening solution
Reagent Type Tissue softener
Form Liquid
Format Bottle
compliance-icons compliance-icons
Product Identifier
  • Cellsoft
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Trimethyl cetyl ammonium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.