missing translation for 'onlineSavingsMsg'
Learn More

Biodesign™ Cellulose Dialysis Tubing Strips

Product Code. 12727476
Change view
Click to view available options
MWCO:
14 kDa
3.5 kDa
8 kDa
Capacity (Metric):
0.32 mL/cm
1.32 mL/cm
1.91 mL/cm
19.13 mL/cm
3.57 mL/cm
6.45 mL/cm
Unit Size:
Pack of 50
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. MWCO Capacity (Metric) unitSize
12727476 14 kDa 1.32 mL/cm Pack of 50
12737476 14 kDa 1.91 mL/cm Pack of 50
12747476 14 kDa 3.57 mL/cm Pack of 50
12757476 14 kDa 6.45 mL/cm Pack of 50
12727486 8 kDa 0.32 mL/cm Pack of 50
12737486 8 kDa 1.91 mL/cm Pack of 50
12747486 8 kDa 3.57 mL/cm Pack of 50
12757486 8 kDa 6.45 mL/cm Pack of 50
12767486 8 kDa 19.13 mL/cm Pack of 50
12707496 3.5 kDa 1.91 mL/cm Pack of 50
12727496 3.5 kDa 6.45 mL/cm Pack of 50
12717496 3.5 kDa 3.57 mL/cm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
This item is not returnable. View return policy
Product Code. 12727476 Supplier Biodesign™ Supplier No. D011
Estimated Shipment: 27-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.

  • Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
  • Just wet in distilled water and use
  • Available in 305mm strips
  • Specially purified for life science research
  • Supplied in an airtight, moisture-proof container for maximum shelf life
  • Available in 14000Da, 8000Da, and 3500Da molecular weight cut offs

Specifications

Product Type Dialysis Tubing Strips
MWCO 14 kDa
Material Cellulose
Diameter (Metric) 6.4 mm
Length (Metric) 30.5 m
Capacity (Metric) 1.32 mL/cm
Color Transparent
Quantity 1 Each
For Use With (Application) For life sciences research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.