missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biodesign™ Cellulose Dialysis Tubing Strips
Description
- Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
- Just wet in distilled water and use
- Available in 305mm strips
- Specially purified for life science research
- Supplied in an airtight, moisture-proof container for maximum shelf life
- Available in 14000Da, 8000Da, and 3500Da molecular weight cut offs
Specifications
Specifications
| Product Type | Dialysis Tubing Strips |
| MWCO | 8 kDa |
| Material | Cellulose |
| Diameter (Metric) | 49.5 mm |
| Length (Metric) | 30.5 m |
| Capacity (Metric) | 19.13 mL/cm |
| Color | Transparent |
| Quantity | 1 Each |
| For Use With (Application) | For life sciences research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?