missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cellulose, microcrystalline

Product Code. 11473318
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473318 500 g 500g
11483318 2500 g 2500g
11493318 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473318 Supplier Thermo Scientific Chemicals Supplier No. A17730.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 9004-34-6

Cellulose, microcrystalline is used as an emulsifier, bulking agent in food production, extender and anti-caking agent. It is also used in vitamin supplements or tablets. It acts as an alternative to carboxymethylcellulose. It finds application in partition chromatography. Further, it is used as a thickener and stabilizer. In addition to this, it is used as an excipient in the pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-34-6
MDL Number MFCD00081512

Specifications

Physical Form Microcrystalline
Merck Index 14,1965
Solubility Information Slightly soluble in sodium hydroxide. Insoluble in water,ethanol,ether and dilute mineral acids.
Quantity 500 g
Chemical Name or Material Cellulose, microcrystalline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.