missing translation for 'onlineSavingsMsg'
Learn More

Cellulose microcrystalline, extra pure, average particle size 90 μm

Product Code. 10198362 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Glass bottle
Plastic drum
Unit Size:
1kg
250g
25kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10198362 1 kg Glass bottle 1kg
10481733 250 g Glass bottle 250g
11916991 5 kg Plastic drum 5kg
15169273 25 kg Fiber Drum 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10198362 Supplier Thermo Scientific Acros Supplier No. 382310010
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 9004-34-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-34-6

Specifications

Color White
pH 5 to 7
Physical Form Fine Crystalline Powder or Granular Powder
Linear Formula (C12H20O10)n
Solubility Information Solubility in water: insoluble.
Quantity 1 kg
Grade Extra Pure
Infrared Spectrum Authentic
Loss on Drying 1.5% max.
Packaging Glass bottle
Particle Size 250 Micron <1% min., 75 Micron >45% min., 32 Micron >70% min.
Residue after Ignition 0.05% max.
Chemical Name or Material Cellulose microcrystalline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.