Learn More
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm High-Flow
Sartorius Stedim Biotech™ Cellulose Nitrate (Mixed Cellulose Ester) Membrane Filters, Gridded, Grey /White 0.45μm High-Flow are recommended for detection of yeasts and molds, particle count, and microscopy.
Brand: Sartorius Stedim Biotech™ 130H6--47----ACR
Additional Details : Weight : 0.50000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Description
- Sterile, individually packaged filters
- Save preparatory time
- Perfect grid printing
- Filter colors allow the best contrast to the colonies or particles
- Outstanding microorganism recovery rate
- High flow membranes
- Sterilized by gamma irradiation
- Compatible with aqueous solutions (pH 4-8), hydrocarbons and several other organic solvents
Specifications
Gray/White Grid | |
0.45 μm | |
130 | |
115 to 145 μm |
Cellulose Nitrate (Cellulose Mixed Ester) | |
Sterile | |
1000/pk. | |
47 mm |
Safety and Handling
- X1000 Cellulose Nitrate Grey - White_ Sterile_ 0,4 5 µm HF
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitrocellulose