Learn More
LABPLAS Cellulose Sani-Sponge with Dey-Engley Neutralizing Broth
Product Code. 17956679
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 100
Product Code. Sterility Unit Size
17956679 Sterile Pack of 100
1 options
Product Code. 17956679 Supplier LABPLAS Supplier No. KSS67115DE
  • ONLINE EXCLUSIVE PRICE
    318.00 €
    258.96 € / Pack of 100
    Save 59.04 €
    18% Off
 Restricted Item
Estimated Shipment: 22-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Cellulose sponge for sampling surfaces, with buffer to neutralize disinfectant.
  • Cellulose Sani-Sponge
  • Saturated with Dey-Engley neutralizing broth
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Gloves included with kit
Specifications
Product Type Hygiene Sponge Sampling Kit
Media Type Neutralizers
Sterility Sterile
Includes Gloves