Learn More
LABPLAS Cellulose Sani-Sponge with Neutralizing Buffer
Product Code. 17926689
Click to view available options
Includes:
Gloves
None
Unit Size:
Pack of 100
Product Code. 17926689
Brand: LABPLAS KSS67110NE
Cellulose sponge for sampling surfaces, with buffer to neutralize disinfectant.
  • Cellulose Sani-Sponge
  • Saturated with neutralizing buffer
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Option to include gloves with kit
Specifications
Product Type Hygiene Sponge Sampling Kit
Media Type Neutralizers
Sterility Sterile
Includes None