Learn More
LABPLAS Cellulose Sani-Stick with Neutralizing Buffer
Product Code. 17946509
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 100
Product Code. Sterility Unit Size
17946509 Sterile Pack of 100
1 options
Product Code. 17946509 Supplier LABPLAS Supplier No. KSS67315NE
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket
Cellulose sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
  • Cellulose Sani-Stick
  • Saturated with neutralizing buffer
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Includes gloves
Specifications
Product Type Hygiene Sponge Sampling Kit
Media Type Neutralizers
Sterility Sterile