missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Centrinone

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
High affinity and selective PLK4 inhibitor (Ki = 0.16 nM). Exhibits >1000-fold selectivity for PLK4 over Aurora A and Aurora B. Depletes centriole and centrosome levels in vitro. Induces cell cycle arrest in normal human cell lines in a p53-dependent manner. Decreases expression of pluripotency markers and induces differentiation in pluripotent stem cells.
Centrinone is a high affinity and selective PLK4 inhibitor (Ki = 0.16 nM). Exhibits >1000-fold selectivity for PLK4 over Aurora A and Aurora B. Depletes centriole and centrosome levels in vitro. Induces cell cycle arrest in normal human cell lines in a p53-dependent manner. Decreases expression of pluripotency markers and induces differentiation in pluripotent stem cells.
Specifications
Specifications
| Chemical Name or Material | 2-[[2-Fluoro-4-[[(2-fluoro-3-nitrophenyl)methyl]sulfonyl]phenyl]thio]-5-methoxy-N-(5-methyl-1H-pyrazol-3-yl)-6-(4-morpholinyl)-4-pyrimidinamine |
| CAS | 1798871-30-3 |
| Quantity | 10 mg |
| Target | Polo-like Kinase Inhibitors |
| Molecular Formula | C26H25F2N7O6S2 |
| Formula Weight | 633.65 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction