Learn More
Labconco™ CentriVap Benchtop Centrifugal Vacuum Concentrator with Acrylic Lid and Heat Boost
BIOPHARMA
Product Code. 15213453 Shop All Labconco Corporation Products
Change view
Click to view available options
Voltage:
0 to 300 V
Unit Size:
Each
Product Code. Voltage Unit Size
15213453 0 to 300 V Each
1 options
Product Code. 15213453 Supplier Labconco™ Supplier No. 7810035
 Restricted Item
Estimated Shipment: 30-03-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
CentriVap Benchtop Vacuum Concentrators are designed to rapidly concentrate multiple small samples using centrifugal force, vacuum and heat. As many as 148 samples may be processed at once. Sample sizes range from a few microliters up to 25 milliliters.
CentriVap Benchtop Vacuum Concentrators are designed to rapidly concentrate multiple small samples using centrifugal force, vacuum and heat. As many as 148 samples may be processed at once. Sample sizes range from a few microliters up to 25 milliliters. CentriVap Concentrators are also available as one component of CentriVap Concentration Systems.
Specifications
Model Benchtop
Includes Acrylic lid and heat boost
Voltage 0 to 300 V
Chamber Material Corrosion Resistant Steel
Plug Type UK