missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Centro Microplate Luminometer LB 963

Product Code. 31777385 Shop All Berthold Technologies Products
Change view
Click to view available options
Model:
Advanced
Aliivibrio fischeri
Research
XS High Sensitivity
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Unit Size
31777385 Advanced Each
31777382 Research Each
31777356 XS High Sensitivity Each
31777364 Aliivibrio fischeri Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 31777385 Supplier Berthold Technologies Supplier No. 7032515
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Easy-to-use microplate luminometer for glow and flash luminescence assays with low background and negligible crosstalk.

The Centro supports all basic luminescence technologies, including:

  • Flash-type Luminescence
  • Glow-type Luminescence
  • Cellular Luminescence
  • Bioluminescence
  • Chemi-Luminescence

Applications

  • Reporter gene assays
  • Aequorin-based calcium monitoring
  • ATP determination
  • Luminescent Immunoassays (LIA, ILMA)
  • DNA probe assays
  • SNP determination
  • CALUX™ Assays

Specifications

Type Luminometer
Display Type No Display
Connection Type USB
Detector Low-noise Photomultiplier Tube in Single Photon Counting Mode
Detection Method Luminescence
Detection Limits <5 amol ATP
Injection Type JET Injectors
Volume (Metric) Dispense 10 to 100 μL
Color Light Gray
For Use With (Application) Luminescence Assays
Voltage 24 VDC
Frequency 50/60 Hz
For Use With (Equipment) 96-well Microplates
Height (Metric) 287 mm
Width (Metric) 334 mm
Depth (Metric) 437 mm
Weight (Metric) 21.8 kg
Includes Luminometer, Power Brick, Accessory Set, User Manual, LightCompass™ Basic Software
System Requirements PC with Windows 10 or Newer
Storage Requirements 0°C to 40°C
Certifications/Compliance CE, NRTL
Model Advanced
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.