missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ CENTRY™ 101 Plate Centrifuge

Product Code. 16348641 Shop All Gilson Products
Change view
Click to view available options
Product Type:
EU-plug
UK-plug
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
16348641 EU-plug Each
16358641 UK-plug -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16348641 Supplier Gilson™ Supplier No. 36117400
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Proprietary swing-out rotor design prevents microplate contents from spilling, increasing the reliability of your microplate reactions. The CENTRY™ 101 Plate Centrifuge accepts popular PCR plates and most microplates.

Decreased spill risk:

  • The CENTRY™ 101 Plate Centrifuge offers a patented swing-out rotor design that keeps the microplate contents from spilling out while concentrating all reaction components in the well bottom, improving the accuracy of your reactions, such as sensitive PCR reactions.
  • The rotor carriers rest at a 75° angle, allowing you to insert plates without sealing tape or caps.
  • At speed, the microplates run vertically.
  • The two-plate capacity CENTRY™ 101 Plate Centrifuge accommodates most PCR and microplates.

Small, but fast:

  • Samples are spun down in less than 20 seconds at 2550 rpm (600 x g).
  • With a small footprint (23 x 26 cm, 9.2 x 10.2 in.), the CENTRY™ 101 Plate Centrifuge easily fits into hoods and on crowded benchtops.
TRUSTED_SUSTAINABILITY

Specifications

Max. Speed 2,550 rpm
Dimensions (L x W x H) Exterior 23 x 26 x 19.7 cm
Frequency 50/60 Hz
Voltage 220/230 V
Product Type EU-plug
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.