missing translation for 'onlineSavingsMsg'
Learn More

Cephalexin Hydrate, TRC

Product Code. 30134242
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
Unit Size:
25g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134242 5 g 5g
30134237 25 g 25g
30010731 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134242 Supplier Toronto Research Chemicals Supplier No. TRCC2568005G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23325-78-2
Molecular Formula C16 H17 N3 O4 S . H2 O
Molecular Weight (g/mol) 365.4
Synonym Cefalexin monohydrate, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-, hydrate (1:1), (6R,7R)-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-(2-amino-2-phenylacetamido)-3-methyl-8-oxo-, monohydrate, D- (8CI), 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[(aminophenylacetyl)amino]-3-methyl-8-oxo-, monohydrate, [6R-[6α,7β(R*)]]-, 5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-[[(2R)-aminophenylacetyl]amino]-3-methyl-8-oxo-, monohydrate, (6R,7R)- (9CI), Cephalexin Asia, Cephalexin monohydrate, Lexine, Pharmexin, Purilex, Rivalexin, (6R,7R)-7-[[(2R)-2-Amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid monohydrate
IUPAC Name (6R,7R)-7-[[(2R)-2-amino-2-phenylacetyl]amino]-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid;hydrate
SMILES O.CC1=C(N2[C@H](SC1)[C@H](NC(=O)[C@H](N)c3ccccc3)C2=O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H17N3O4S.H2O/c1-8-7-24-15-11(14(21)19(15)12(8)16(22)23)18-13(20)10(17)9-5-3-2-4-6-9;/h2-6,10-11,15H,7,17H2,1H3,(H,18,20)(H,22,23);1H2/t10-,11-,15-;/m1./s1
Formula Weight 365.1045
Percent Purity >95%
Chemical Name or Material Cefalexin monohydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.