missing translation for 'onlineSavingsMsg'
Learn More

Cephaloridine Monohydrate, TRC

Product Code. 30138125
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
Unit Size:
250mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138125 250 mg 250mg
30135807 50 mg 50mg
30005534 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138125 Supplier Toronto Research Chemicals Supplier No. TRCC258600250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102039-86-1
Molecular Formula C19H17N3O4S2•H2O
Molecular Weight (g/mol) 415.49
Synonym 1-[[(6R,7R)-2-Carboxy-8-oxo-7-[[2-(2-thienyl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl]-pyridinium Inner Salt Hydrate;
IUPAC Name (6R,7R)-8-oxo-3-(pyridin-1-ium-1-ylmethyl)-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate;hydrate
SMILES Cc1c2c(CCC2)c(cc1)C(=O)c3c4c(cccc4)ccc3

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H17N3O4S2.H2O/c23-14(9-13-5-4-8-27-13)20-15-17(24)22-16(19(25)26)12(11-28-18(15)22)10-21-6-2-1-3-7-21;/h1-8,15,18H,9-11H2,(H-,20,23,25,26);1H2/t15-,18-;/m1./s1
Formula Weight 433.0766
Percent Purity >95%
Chemical Name or Material Cephaloridine Monohydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.