missing translation for 'onlineSavingsMsg'
Learn More

Cephalosporin C zinc salt, MedChemExpress

Product Code. 16498688
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498688 10 mg 10mg
16464577 50 mg 50mg
16418726 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498688 Supplier Medchem Express Supplier No. HYB1299A
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Cephalosporin C zinc salt is a potent inhibitor of SAMHD1 with an IC50 of 1.1 μM. Cephalosporin C zinc salt also has moderate anti-orthopoxvirus activity.

Cephalosporin C zinc salt is a potent inhibitor of SAMHD1 with an IC50 of 1.1 μM. Cephalosporin C zinc salt also has moderate anti-orthopoxvirus activity.

Specifications

Chemical Name or Material Cephalosporin C zinc salt
CAS 59143-60-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C16H19N3O8SZn
Quantity 10 mg
Solubility Information DMSO : 5 mg/mL (10.44 mM; ultrasonic and adjust pH to 3 with 1M HCl)
SMILES CC(OCC1=C(C([O-])=O)N(C([C@H]2NC(CCC[C@@H](N)C([O-])=O)=O)=O)[C@]2([H])SC1)=O.[Zn+2]
Molecular Weight (g/mol) 478.78
Formula Weight 478.78
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.