missing translation for 'onlineSavingsMsg'
Learn More

Cephalosporin C Zinc Salt, TRC

Product Code. 30008228
Change view
Click to view available options
Quantity:
5 mg
50 mg
100 mg
Unit Size:
100mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008228 5 mg 5mg
30008119 100 mg 100mg
30004046 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008228 Supplier Toronto Research Chemicals Supplier No. TRCC2587505MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59143-60-1
Molecular Formula C16 H19 N3 O8 S Zn
Molecular Weight (g/mol) 478.81
Synonym (6R,7R)-3-[(Acetyloxy)methyl]-7-[[(5R)-5-amino-5-carboxy-1-oxopentyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid Zinc Salt, , [6R-[6α,7β(R*)]]-3-[(Acetyloxy)methyl]-7-[(5-amino-5-carboxy-1-oxopentyl)amino]-8-oxo-5-Thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid Zinc Salt
IUPAC Name (6R,7R)-3-(acetyloxymethyl)-7-[[(5R)-5-amino-5-carboxypentanoyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
SMILES CC(=O)OCC1=C2N3[C@H](SC1)[C@H](NC(=O)CCC[C@@H](N)C(=O)O[Zn]OC2=O)C3=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H21N3O8S.Zn/c1-7(20)27-5-8-6-28-14-11(13(22)19(14)12(8)16(25)26)18-10(21)4-2-3-9(17)15(23)24;/h9,11,14H,2-6,17H2,1H3,(H,18,21)(H,23,24)(H,25,26);/q;+2/p-2/t9-,11-,14-;/m1./s1
Formula Weight 477.0184
Chemical Name or Material Cephalosporin C Zinc Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.