missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Ceramus™ Classic Fix
A universal bottle attachment dispenser for liquid handling.
Brand: Hirschmann™ 9311100
Additional Details : Weight : 0.25000kg
Description
- Universal bottle attachment dispenser with fixed volume configuration
- Certified conformity
- With ceramic piston
- Suitable for steam sterilization up to 121°C
- Delivery complete with A 32 mm screw thread, 3 threaded adapters (A 28, S 40, A 45), 1 suction hose with union nut, 1 ejection unit and 1 fitting wrench
- With individual serial number and calibration certificate
Specifications
0.6% | |
Bottle Attachment Dispenser | |
Screw thread, 3 threaded adapters, 1 suction hose, 1 ejection unit, 1 fitting wrench |
0.2% | |
1 mL |