missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Ceramus™ Fix Bottle Top Dispensers

Product Code. 10548062
Change view
Click to view available options
Volume (Metric):
10 mL
50 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10548062 50 mL Each
10267910 10 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10548062 Supplier Hirschmann™ Supplier No. 9362100
  • ONLINE EXCLUSIVE PRICE
    759.15 €
    545.00 € / Pack of 1
    Save 214.15 €
    28% Off
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Guarantees consistent precision and volume dispensing with reproducible results independent of the user. Hirschmann™ Ceramus™ Classic Dispensers are the proven unit for manual dispensing, featuring superior-quality materials, simple handling and well-engineered user comfort to make laboratory routines easier.

  • For fixed and variable volumes, from 0.2mL to 60mL
  • Complete unit can be steam sterilized at 121°C without dismantling
  • Free rotation
  • Media recirculating system for simple and rapid venting without reagent loss
  • Digital volume configuration engagement, calibrated for a lifetime
  • Use within a temperature range of 4°C to 50°C
  • Individual quality certificate with conformity certificate

Specifications

Product Type Bottletop Dispenser
Material Ceramic
Volume (Metric) 50 mL
For Use With (Equipment) Bottle
Includes 3 Threaded adapters (A 32, S 40, A 38), 1 suction hose with union nut, 1 ejection unit and 1 fitting wrench, with individual serial number and calibration certificate.
Precision 0.6%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.