missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cerberus 1 Goat anti-Mouse, Polyclonal, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF1986-SP
This item is not returnable.
View return policy
Description
Cerberus 1 Polyclonal specifically detects Cerberus 1 in Mouse samples. It is validated for Western Blot.Specifications
| Cerberus 1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CER1, cerberus, cerberus 1 (Xenopus laevis) homolog (cysteine knot superfamily), cerberus 1, cysteine knot superfamily, homolog (Xenopus laevis), cerberus-related 1, Cerberus-related protein, DAN domain family member 4, DAND4, MGC119894, MGC119895, MGC96951 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9350 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| O55233 | |
| CER1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cerberus 1 Asp41-Pro272 Accession # O55233 | |
| 25 μg | |
| Primary | |
| Detects mouse Cerberus 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Cerberus is observed. | |
| Mouse | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?