missing translation for 'onlineSavingsMsg'
Learn More

Ceritinib, MedChemExpress

Product Code. 16481186
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
Unit Size:
100mg
10mg
1g
200mg
2g
500mg
50mg
5mg
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481186 200 mg 200mg
16471776 500 mg 500mg
16471539 1 g 1g
16442807 50 mg 50mg
16439277 5 mg 5mg
16436866 100 mg 100mg
16426728 10 mg 10mg
16420256 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 16481186 Supplier Medchem Express Supplier No. HY15656
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ceritinib (LDK378) is a selective, orally bioavailable, and ATP-competitive ALK tyrosine kinase inhibitor with an IC50 of 200 pM. Ceritinib (LDK378) also inhibits IGF-1R, InsR, and STK22D with IC50 values of 8, 7, and 23 nM, respectively. Ceritinib (LDK378) shows great antitumor potency.

Ceritinib (LDK378) is a selective, orally bioavailable, and ATP-competitive ALK tyrosine kinase inhibitor with an IC50 of 200 pM. Ceritinib (LDK378) also inhibits IGF-1R, InsR, and STK22D with IC50 values of 8, 7, and 23 nM, respectively. Ceritinib (LDK378) shows great antitumor potency.

Specifications

Chemical Name or Material Ceritinib
CAS 1032900-25-6
Color White
Purity Grade Notes Research
Molecular Formula C28H36ClN5O3S
Quantity 200 mg
Synonym LDK378
Solubility Information DMSO : 5.6 mg/mL (10.03 mM; Need ultrasonic) ∣Ethanol : ≥ 3.33 mg/mL (5.97 mM)
SMILES CC(C)OC1=CC(C2CCNCC2)=C(C)C=C1NC3=NC=C(Cl)C(NC4=CC=CC=C4S(=O)(C(C)C)=O)=N3
Molecular Weight (g/mol) 558.14
Formula Weight 558.14
Percent Purity 97.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.