missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific™

86.60 € - 390.98 €

Chemical Identifiers

CAS 14457-87-5
MDL Number MFCD00016004
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15427465
SDS View Documents
Thermo Scientific Alfa Aesar
047194.03
1g
ONLINE EXCLUSIVE PRICE
88.30 €
86.60 € / 1g
Save 1.70 €
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15437465
SDS View Documents
Thermo Scientific Alfa Aesar
047194.06
5g
ONLINE EXCLUSIVE PRICE
175.00 €
172.00 € / 5g
Save 3.00 €
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15447465
SDS View Documents
Thermo Scientific Alfa Aesar
047194.14
ONLINE EXCLUSIVE PRICE
399.00 €
390.98 € / 25g
Save 8.02 €
2% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
Specifications

Chemical Identifiers

14457-87-5
MFCD00016004

Specifications

14457-87-5
14,1995
379.83
Odorless
Hygroscopic
(metals basis)
MFCD00016004
Soluble in water.
99.99%
Ampule
5.18 g/mL
Cerium(III) bromide, Ultra dry
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 238-447-0

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.