missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) chloride, 99.9%, pure, anhydrous

Product Code. 10650893 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10650893

Brand: Thermo Scientific Acros 369750250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7790-86-5 | CeCl3 | 246.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-86-5
Molecular Formula CeCl3
Molecular Weight (g/mol) 246.48
MDL Number MFCD00010929
InChI Key VYLVYHXQOHJDJL-UHFFFAOYSA-K
Synonym cerium iii chloride, cerous chloride, cerium iii chloride, anhydrous, cerium-144 chloride, cerium chloride generic, cerium-chloride, cerium chloride octahydrate, cerous iii-chloride
PubChem CID 24636
ChEBI CHEBI:35458
IUPAC Name cerium(3+);trichloride
SMILES [Cl-].[Cl-].[Cl-].[Ce+3]

Specifications

Boiling Point 1727.0°C
Melting Point 848.0°C
Color White to Gray
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.90%
Linear Formula CeCl3
Merck Index 15, 1996
Solubility Information Solubility in water: soluble
Formula Weight 246.48
Percent Purity 99.9%
Grade Pure
Packaging Glass bottle
Chemical Name or Material Cerium(III) chloride, Anhdrous
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cerium(III) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informazioni supplementari
  • MIXTURE LIST-Contains : Cerium chloride

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato