missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) chloride heptahydrate, 99%

Product Code. 10732831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
Product Code. Quantity Packaging unitSize
10732831 500 g Glass bottle 500g
10688124 25 g Glass bottle 25g
10082710 100 g Glass bottle 100g
3 options
This item is not returnable. View return policy

Product Code. 10732831

Brand: Thermo Scientific Acros 211825000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18618-55-8 | CeCl3·7H2O | 372.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18618-55-8
Molecular Formula CeCl3·7H2O
Molecular Weight (g/mol) 372.58
MDL Number MFCD00149634

Specifications

Melting Point 848.0°C
Color Yellow
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula CeCl3·7H2O
Fieser 01,121; 09,99; 15,72; 16,4; 17,314
Merck Index 10, 1956
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol
Formula Weight 372.58
Percent Purity 99%
Packaging Glass bottle
Density 3.97
Chemical Name or Material Cerium(III) chloride heptahydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.